Avantor® ACE® Excel® NH₂, HPLC/UHPLC Columns, Analytical, 1.7 µm
Supplier: Avantor
EXL-1714-0502U EXL-1714-0503U EXL-1714-1002U EXL-1714-1003U EXL-1714-7503U
76394-516EA 6059.41 CAD
76394-516 76394-518 76394-520 76394-522 76394-524 76394-526 76394-528 76394-530 76394-532 76394-534 76394-536 76394-538
Avantor® ACE® Excel® NH₂, HPLC/UHPLC Columns, Analytical, 1.7 µm
Chromatography Columns
These columns with proprietary bonding technology provide high robustness with excellent column lifetimes.
  • Usable in reversed phase, normal phase, weak anion-exchange and HILIC modes
  • Ideal for the analysis of sugars
  • Ultra-inert, high efficiency 1.7, 3 and 5 µm particles for optimum peak shape
  • Method validation kits contain three different batches of the same phase - available on request
These columns are made of stainless steel.