Avantor® ACE® CN-ES, HPLC Columns, Microbore, 3 µm
Supplier: Avantor
ACE-11130102GD ACE-11130103GD
76384-426PK 505.16 CAD
76384-426 76384-428 76384-430 76384-432 76384-434 76384-436 76384-438 76384-440 76384-442 76384-444 76384-446 76384-448 76384-450 76384-452 76384-454 76384-456 76384-458 76384-460 76384-462 76384-464 76384-466 76384-468 76384-488 76384-490 76384-492 76384-494
Avantor® ACE® CN-ES, HPLC Columns, Microbore, 3 µm
Chromatography Columns
Avantor® ACE® CN-ES combines CN polar selectivity with enhanced hydrophobicity, enabling the benefits of both interactions to be fully exploited. Extra phase stability and increased column lifetime is provided by an extended alkyl chain spacer between the silica surface and cyano group. Avantor® ACE® CN-ES is suitable for both reversed phase and normal phase separations.
  • Ultra high purity, base deactivated silica with exceptional reproducibility
  • With hydrophobic plus CN polar selectivity, especially useful for polar molecules
  • Wide range of particle sizes (1.7 to 10 µm) and complementary phases
  • Method validation kits contain three different batches of the same phase - available on request
These stainless steel columns are available in a wide range of dimensions from capillary to preparative for HPLC and UHPLC separations.