Avantor® ACE® CN-ES, HPLC Columns, 5 µm
Supplier: Avantor
ACE-1213-1510 ACE-1213-2510 ACE-1213-2520 ACE12130110GD ACE-1213-1030
76386-420EA 6061.37 CAD
76386-420 76386-422 76386-424 76386-426 76386-428 76386-430 76386-432 76386-434 76386-436 76386-438 76386-440 76386-442 76386-444 76386-446 76386-448 76386-450 76386-452 76386-454 76386-356 76386-358 76386-364 76386-366 76386-368 76386-370 76386-376 76386-378 76386-380 76386-382 76386-384 76386-386 76386-388 76386-390 76386-392 76386-394 76386-396 76386-398 76386-400 76386-402 76386-404 76386-406 76386-408 76386-410 76386-412 76386-414 76386-470 76386-472 76386-474 76386-476 76386-482 76386-484
Avantor® ACE® CN-ES, HPLC Columns, 5 µm
Chromatography Columns
Avantor® ACE® CN-ES combines CN polar selectivity with enhanced hydrophobicity, enabling the benefits of both interactions to be fully exploited. Extra phase stability and increased column lifetime is provided by an extended alkyl chain spacer between the silica surface and cyano group. Avantor® ACE® CN-ES is suitable for both reversed phase and normal phase separations.
  • Ultra high purity, base deactivated silica with exceptional reproducibility
  • With hydrophobic plus CN polar selectivity, especially useful for polar molecules
  • Wide range of particle sizes (1.7 to 10 µm) and complementary phases
  • Method validation kits contain three different batches of the same phase - available on request
These stainless steel columns are available in a wide range of dimensions from capillary to preparative for HPLC and UHPLC separations.