Avantor® ACE® C18, HPLC Columns, 2 µm, Microbore
Supplier: Avantor
ACE-101-05005S ACE-101-15005
76383-584EA 1737.78 CAD
76383-584 76383-690 76383-692 76383-694 76383-696 76383-498 76383-500 76383-502 76383-504 76383-506 76383-508 76383-510 76383-512 76383-522 76383-524 76383-526 76383-528 76383-570 76383-572 76383-574 76383-576 76383-578 76383-580 76383-582
Avantor® ACE® C18, HPLC Columns, 2 µm, Microbore
Chromatography Columns
Avantor® ACE® C18 columns provide great reproducibility and column lifetime. These stainless steel columns are available in a wide range of particle sizes and dimensions from capillary to preparative. These columns are designed for a wide range of chromatographic applications, to provide great performance with acidic, basic and neutral molecules.
  • Ultra high purity, base deactivated silica with exceptional reproducibility
  • Ideal for a wide range of analytes differing in hydrophobicity
  • Wide range of particle sizes (2 to 10 µm) and complementary phases