Avantor® ACE® Excel® Silica, HPLC/UHPLC Columns, Analytical, 2 µm
Supplier: Avantor
ACE-1P7-GD2U ACE-1P7-GD4U
76392-946EA 1326.17 CAD
76392-946 76392-948 76392-950 76392-952 76392-954 76392-956 76392-958 76392-960 76392-962 76392-964 76392-966 76392-968 76392-970 76392-972 76392-974 76392-976 76392-978 76392-980 76392-982 76392-984 76392-986 76392-988 76392-990 76392-992 76408-434 76408-436
Avantor® ACE® Excel® Silica, HPLC/UHPLC Columns, Analytical, 2 µm
Chromatography Columns
Avantor® ACE® Excel® Silica columns provide great reproducibility and column lifetime. These stainless steel columns are available in a wide range of particle sizes and dimensions from capillary to preparative.
  • Ultra high purity, base deactivated silica with exceptional reproducibility
  • Wide range of particle sizes (2 to 5 µm) and complementary phases
  • Method validation kits contain three different batches of the same phase - available on request
These columns are designed for a wide range of chromatographic applications, to provide great performanceMore Product Information