Avantor® ACE® Excel® Phenyl, HPLC/UHPLC Columns, Analytical, 2 µm
Supplier: Avantor
EXL-105-0502U EXL-105-0503U EXL-105-0546U EXL-105-1002U EXL-105-1003U EXL-105-1046U EXL-105-1202U EXL-105-1246U EXL-105-1502U EXL-105-1503U EXL-105-1546U EXL-105-7502U EXL-105-7503U EXL-105-7546U ACE-1P5-GD4U
76392-850EA 1404.86 CAD
76392-850 76392-852 76392-854 76392-856 76392-858 76392-860 76392-862 76392-864 76392-866 76392-868 76392-870 76392-872 76392-874 76392-876 76392-878 76392-880 76392-882 76392-884 76392-886 76392-888 76392-890 76392-892 76392-894 76392-896 76408-426 76408-428
Avantor® ACE® Excel® Phenyl, HPLC/UHPLC Columns, Analytical, 2 µm
Chromatography Columns
These Avantor® ACE® Excel® Phenyl columns provide great reproducibility and column lifetime. These stainless steel columns are available in a wide range of particle sizes and dimensions from capillary to preparative.
  • Ultra high purity, base deactivated silica with exceptional reproducibility
  • Offers strong π-π interactions
  • Ideal for analytes with π-bonding, electron withdrawing or delocalised groups
  • Wide range of particle sizes available (2, 3 and 5 µm) and complementary phases
  • Method validation kits contain three different batches of the same phase -
More Product Information