Avantor® ACE®, HPLC/UHPLC Columns, Extended Method Development Kits, 1.7 µm
Supplier: Avantor
MDKE-17-0502U MDKE-17-0503U MDKE-17-1002U MDKE-17-1003U
76396-804EA 2759.63 CAD
76396-804 76396-806 76396-808 76396-810
Avantor® ACE®, HPLC/UHPLC Columns, Extended Method Development Kits, 1.7 µm
Chromatography Columns
Avantor® ACE® Method Development Kits (MDK) are designed to maximise selectivity, offering a powerful and reliable approach to UHPLC/HPLC method development. Based upon an ultra-inert, high efficiency silica, Avantor® ACE® phases incorporate the latest developments in LC stationary phase design, providing chromatographers with more choices for alternative selectivity, without compromising stability or robustness.
  • Porous, solid core, HILIC and biomolecule kit options
  • Wide range of particle sizes (1.7 and 5 µm ) and complementary phases
  • Analytical IDs in a
More Product Information