Avantor® Partisil ODS3, HPLC Columns
Supplier: Avantor
P10ODS3-10C5 P10ODS3-150A P10ODS3-250A P10ODS3-300AF P5ODS3-100A P5ODS3-10C5 P5C8-125A P5ODS3-150A P5ODS3-250A
76401-854EA 2376.18 CAD
76401-854 76401-856 76401-858 76401-860 76401-862 76401-864 76401-866 76401-868 76401-870 76401-872 76401-874 76401-876 76401-878 76401-880 76401-882 76401-884 76401-886 76401-888 76401-890 76401-892 76401-894 76401-896 76402-104 76402-106 76402-108 76402-110 76402-112 76402-114 76402-116 76402-118 76402-120 76402-122 76402-124 76402-126 76402-128 76402-130 76402-132 76402-134 76402-136 76402-138 76402-140 76402-142 76402-144 76402-146 76402-148
Avantor® Partisil ODS3, HPLC Columns
Chromatography Columns
Avantor® Partisil® was one of the first commercially available irregular silicas with a large surface area giving it a high loading capacity.
  • Easy to use
  • Irregular porous silica
Avantor® Partisil® ODS3 is an intermediate carbon load C18 phase used for pharmaceutical, natural products, food, biological and environmental pollutants applications.
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification ISO 14001:2015 (530840 UM15)