LiChrospher® RP-18E, HPLC Columns
Supplier: Avantor
LISPRP18E510C5 LISPRP18E5125F LISPRP18E5150A LISPRP18E5150M LISPRP18E5150S LISPRP18E5200A LISPRP18E5250A LISPRP18E550AF LISRP18E10250F SPRP18E510C5SK
76400-598EA 1355.66 CAD
76400-598 76400-600 76400-602 76400-604 76400-606 76400-608 76400-610 76400-612 76400-614 76400-616 76400-618 76400-620 76400-622 76400-624 76400-626 76400-628 76400-630 76400-632 76400-634 76400-636 76400-638 76400-640 76400-642 76400-644 76400-646 76400-768 76400-770 76402-352
LiChrospher® RP-18E, HPLC Columns
Chromatography Columns
These LiChrospher® columns are silica available both end capped and non-end capped C18 phases. This feature enhances the chromatography selectivity options.
  • Spherical porous silica
  • High efficiency packed columns
  • Stainless steel columns