Avantor® Ultrasphere® Silica (Si), HPLC Columns
Supplier: Avantor
235341 235342 237574
76398-096EA 4550.42 CAD
76398-096 76398-098 76398-100 76398-106 76398-108 76398-128 76398-162
Avantor® Ultrasphere® Silica (Si), HPLC Columns
Chromatography Columns
Avantor® Ultrasphere® columns remain widely referenced within both industry and academia and are recognized to provide excellent chromatographic performance.
  • 3 and 5 μm spherical porous silica
  • High reproducibility, narrow particle size distribution
  • Optimal surface coverage for long column lifetime