Avantor® Ultrasphere® Cyano (CN), HPLC Columns
Supplier: HICHROM LIMITED
237573 244070 244071
76398-110EA 1638.84 CAD
76398-110 76398-112 76398-126 76398-168 76398-170 76398-172 76398-174
Avantor® Ultrasphere® Cyano (CN), HPLC Columns
Chromatography Columns
Avantor® Ultrasphere® columns remain widely referenced within both industry and academia and are recognized to provide excellent chromatographic performance.
  • 3 and 5 μm spherical porous silica
  • High reproducibility, narrow particle size distribution
  • Optimal surface coverage for long column lifetime