Avantor® Ultrasphere® Octyl (C8), HPLC Columns
Supplier: HICHROM LIMITED
235332 235333
76398-086EA 3865.08 CAD
76398-086 76398-088 76398-090 76398-114 76398-116 76398-156 76398-166 76398-176
Avantor® Ultrasphere® Octyl (C8), HPLC Columns
Chromatography Columns
Avantor® Ultrasphere® columns remain widely referenced within both industry and academia and are recognized to provide excellent chromatographic performance.
  • 3 and 5 μm spherical porous silica
  • High reproducibility, narrow particle size distribution
  • Optimal surface coverage for long column lifetime