Avantor® Partisil Silica, HPLC Columns
Supplier: Avantor
P10-150AF P10-250A P5-100A P5-150A P5-200A P5-250A P5-250AF P5-250AS
76401-690EA 1573.62 CAD
76401-690 76401-692 76401-694 76401-698 76401-700 76401-702 76401-704 76401-706 76401-708 76401-710 76401-712 76401-714 76401-716 76401-718 76401-720 76401-722 76401-724 76401-726 76401-728 76402-022 76402-024 76402-026 76402-028 76402-030 76402-032 76402-034 76402-036 76402-038 76402-040 76402-042 76402-044 76402-046 76402-048 76402-050 76402-052 76402-054 76402-056 76402-058 76402-060
Avantor® Partisil Silica, HPLC Columns
Chromatography Columns
Avantor® Partisil® was one of the first commercially available irregular silicas.
  • Irregular porous silica
  • 5 and 10 µm particle sizes
  • Hichrom high efficiency
A large surface area gives it a high loading capacity. Avantor® Partisil® ion-exchange materials are widely referenced and remain one of the most popular choices for analysts.