Avantor® Partisil SCX, HPLC Columns
Supplier: Avantor
P10SCX-10C5 P10SCX-10C5-SK P10SCX-150A P10SCX-250A P10SCX-250AF P5SCX-100A P5SCX-100AM P5SCX-10C5 P5SCX-10C5-SK P5SCX-10CM5 P5SCX-125A P5SCX-250A P5SCX-250AF P5SCX-50A
76401-976EA 1573.62 CAD
76401-976 76401-978 76401-980 76401-982 76401-984 76401-986 76401-988 76401-990 76401-992 76401-994 76401-996 76401-998 76402-000 76402-002 76402-004 76402-006 76402-008 76402-010 76402-012 76402-014 76402-016 76402-018 76402-020 76402-232 76402-234 76402-236 76402-238 76402-240 76402-242 76402-244 76402-246 76402-248 76402-250 76402-252 76402-254 76402-256 76402-258 76402-260 76402-262 76402-264 76402-266 76402-268 76402-270 76402-272
Avantor® Partisil SCX, HPLC Columns
Chromatography Columns
Avantor® Partisil® was one of the first commercially available irregular silicas with a large surface area giving it a high loading capacity.
  • Easy to use
  • Irregular porous silica
Avantor® Partisil® SCX is a strong cation exchange phase based on benzenesulphonic acid groups and is stable over the pH range 1.5 to 7.0. At intermediate pH, use of a Solvecon eluent conditioning column is recommended to further enhance column lifetime. Suitable for use with nucleosides, amino acids, polyamines, drugs and other cationic species.
Column hardware: Stainless steel.
Caution: * Use with freestanding holder and column coupler, please see 'HPLC Guard Column Holders and Accessories'.