LiChrospher® Diol, HPLC Columns
Supplier: HICHROM LIMITED
LISPOH-5-100AF LISPOH-5-10C5 LISPOH-5-10CE5 LISPOH-5-10CM5 LISPOH-5-125AF LISPOH-5-150AM LISPOH-5-250A LISPOH-5-250AF LISPOH-5-250AM LISPOH510C5SK
76400-488EA 1355.62 CAD
76400-488 76400-490 76400-492 76400-494 76400-496 76400-498 76400-500 76400-502 76400-504 76400-506 76400-508 76400-510 76400-512 76400-514 76400-516 76400-518 76400-520 76400-522 76400-524 76400-526 76400-528 76400-530 76400-532 76400-534 76400-536 76400-538 76400-540
LiChrospher® Diol, HPLC Columns
Chromatography Columns
LiChrospher® is a silica available in both endcapped and non-endcapped C8 and C18 phases.
  • Spherical porous silica
  • Base deactivated LiChrospher® RP-select B
  • High efficiency packed columns
Column hardware: Stainless steel.