LiChrospher® Si 100, HPLC Columns
Supplier: Avantor
LISP100-5-10C5 LISP100-5-150A LISP100-5-250A
76400-310EA 1290.88 CAD
76400-310 76400-312 76400-314 76400-316 76400-318 76400-320 76400-322 76400-324 76400-326 76400-328 76400-330
LiChrospher® Si 100, HPLC Columns
Chromatography Columns
LiChrospher® is a silica available in both endcapped and non-endcapped C8 and C18 phases.
  • Spherical porous silica
  • Base deactivated LiChrospher® RP-select B
  • High efficiency packed columns
Column hardware: Stainless steel.