Avantor® Partisil C8, HPLC Columns
Supplier: Avantor
4222-232 4229-001 4239-001 P10C8-150AS P10C8-250A P5C8-100A P5C8-100AF P5C8-10C5 P5C8-10C5-SK P5C8-150AS
76398-232EA 2066.09 CAD
76398-232 76398-238 76398-260 76398-346 76401-730 76401-732 76401-734 76401-736 76401-738 76401-740 76401-742 76401-744 76401-746 76401-748 76401-750 76401-752 76401-754 76401-756 76401-758 76401-760 76401-762 76401-764 76401-766 76402-062 76402-064 76402-066 76402-068 76402-070 76402-072 76402-074 76402-076 76402-078 76402-080 76402-082 76402-084 76402-086 76402-088 76402-090 76402-092 76402-094 76402-096 76402-098 76402-100 76402-102
Avantor® Partisil C8, HPLC Columns
Chromatography Columns
Avantor® Partisil® was one of the first commercially available irregular silicas with a large surface area giving it a high loading capacity.
  • Easy to use
  • Irregular porous silica
  • Stainless steel column hardware
Avantor® Partisil® C8 is recommended for ion-pair chromatography.
Caution: * Use with freestanding holder HI-161 and column coupler HI-081, please see 'HPLC Guard Column Holders and Accessories'.
Order Now
SPECIFICATIONS
Environmentally Preferable