LiChrospher® Si 60, HPLC Columns
Supplier: Avantor
LISP60-5-10CP3 LISP60-5-125AF LISP60-5-150A LISP60-5-250A
76400-334EA 1290.81 CAD
76400-334 76400-336 76400-338 76400-340 76400-342 76400-344 76400-346 76400-348 76400-350 76400-352 76400-354 76400-356 76400-358 76400-360 76400-362 76400-364 76400-366 76400-368 76400-370 76400-372 76400-374 76400-376 76400-378 76400-380 76400-382 76400-386
LiChrospher® Si 60, HPLC Columns
Chromatography Columns
LiChrospher® is a silica available in both endcapped and non-endcapped C8 and C18 phases.
  • Spherical porous silica
  • Base deactivated LiChrospher® RP-select B
  • High efficiency packed columns
Column hardware: Stainless steel.