VeriSpec® ICP Multi-Element Standard 4, RICCA Chemical Company
Supplier: Ricca Chemical
RV010706-100N RV010706-500N
RCRV010706100NEA 933.78 CAD
RCRV010706100N RCRV010706500N
VeriSpec® ICP Multi-Element Standard 4, RICCA Chemical Company
Standards Cationic Standards Multi-Element Standards
4 component standard.
5000 ppm: Ca, K, Mg, Na
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility