Avantor® Hichrom, Chromatography Tubing, Stainless Steel
Supplier: HICHROM LIMITED
01-SS-05-3M 01-SS-07-10M 01-SS-07-3M 01-SS-10-1.5M 01-SS-10-10M 01-SS-10-3M 01-SS-20-10M 01-SS-20-3M 01-SS-30-1.5M 01-SS-30-10M 01-SS-30-5M 01-SS-40-1.5M 01-SS-40-10M 01-SS-40-3M
76397-920EA 437.42 CAD
76397-920 76397-922 76397-924 76397-926 76397-928 76397-930 76397-932 76397-934 76397-936 76397-938 76397-940 76397-942 76397-944 76397-946 76397-948 76397-950 76397-952
Avantor® Hichrom, Chromatography Tubing, Stainless Steel
Tubing Chromatography Tubing
Tubings are madeup of 316 Stainless steel, precision bore ¹/₁₆" type tubing.
  • Operating temperature - no limit
  • May corrode with chloride salt or other buffers
  • May adsorb proteins
These 0.005 and 0.007" Ø - suitable for minimal dead volume injector-column or column-detector connections of microbore or short columns containing 3 µm material. Sample and material must be filtered to avoid blockage.
The tube with 0.10' Ø - suitable for connection of analytical and semi-preparative columns containing 5 and 10 µm material. It should be used in all parts of the system where dead volume needs to be kept to minimum.
These 0.20, 0.30 and 0.40" Ø - suitable for use in preparative systems or in non-critical connections (pump to sample injector where low flow resistance is more important than low internal volume).