Flip-Top Hinged Cardboard Freezer Boxes
Supplier: MTC BIO, INC.
R2700 R2700-A R2700-B R2700-G R2700-O R2700-P R2700-R R2781 R2781-A R2781-B R2781-G R2781-O R2781-P R2781-R
470324-968PK 47.55 CAD
470324-968 470324-970 470324-972 470324-974 470324-976 470324-978 470324-980 470324-982 470324-984 470324-986 470324-988 470324-990 470324-992 470324-994
Flip-Top Hinged Cardboard Freezer Boxes
Boxes Cryoboxes
Our hinged lid boxes are made of recyclable cardboard, with a liquid resistant outer coating.
  • Accommodates 1.5 and 2.0 ml tubes, as well as most 2.0 ml cryogenic vials
  • Alpha-numeric coding helps sample identification
  • Standard sized boxes will fit in your existing racks
  • Available in six colors, in two different tube configurations
Choose between 81 or 100-place cardboard dividers. Each box has drain slots on the bottom surface and numbered grid markings in almost every conceivable location (top, bottom, sides, and so on).
Ordering information: Available in white and five individual colors, plus a 'Rainbow Pack'. Boxes are shipped fully assembled.