Avantor® Hichrom, Cutters, Polymeric Tubing
Supplier: Avantor
HI-194 HI-197
76399-194EA 79.27 CAD
76399-194 76399-198
Avantor® Hichrom, Cutters, Polymeric Tubing
Cutters
The Clean-cut™ cutter is designed specifically for cutting polymeric HPLC tubing from ¹/₁₆ to ¹/₈" with no distortion of cross-section and without closing the bore.
  • Cuts PEEK and PTFE tubing
  • Square, burr-free cuts
  • Convenient, replaceable blade
The 'Guillotine' cutter is an alternative design for cutting polymeric tubing of ¹/₁₆ to ¹/₂". A razor-sharp blade cuts while a V-shaped grove on the handle holds the tubing at the right angle to produce a square cut.