Avantor® Hichrom PEEKsil™, Tubing
Supplier: VWR
Avantor® Hichrom
HI-1083 HI-320
76399-354PK 175.44 CAD
76399-354 76399-056
Avantor® Hichrom PEEKsil™, Tubing
Tubing Chromatography Tubing
PEEKsil™ tubing is mechanically strong and has ideal characteristics for sealing with polymer fittings. It is comprised of high quality fused silica sheathed by PEEK tubing giving it excellent chemical compatibility.