Avantor® Hichrom PEEK™, Unions, Fittings
Supplier: Avantor
HI-055 HI-059
76399-026EA 166.72 CAD
76399-026 76399-030
Avantor® Hichrom PEEK™, Unions, Fittings
Fittings and Connectors
Fitings that are chemically inert and biocompatible
  • Zero dead volume connection
  • Pressure rated to 344 bar
  • Complete with two 76399-380 fingertight fitting
Swept volume: 0.07 µl and 0.28 µl.
Order Now
SPECIFICATIONS
Environmentally Preferable