Avantor® Hichrom Bottom-of-the-Bottle™, Inlet Solvent Filters
Supplier: Avantor
HI-579 HI-580
76399-404EA 163.93 CAD
76399-404 76399-406
Avantor® Hichrom Bottom-of-the-Bottle™, Inlet Solvent Filters
Filters Liquid Process Filters
These filters are made from 100% PEEK.
  • Biocompatible filters
  • Two built-in PEEK frits
The bottom frit (2 or 10 µm) will draw solvents from within 2 mm of the bottom of the solvent bottle. The 2 μm frit on the side may be used for a 203 mm Ext.Ø helium sparging line.