Avantor® Hichrom, Inlet Solvent Filters
Supplier: Avantor
HI-583 HI-584
76399-410EA 120.74 CAD
76399-410 76399-412
Avantor® Hichrom, Inlet Solvent Filters
Filters Liquid Process Filters
These solvent inlet filters are used at the low-pressure inlet side of the pump to help protect check valves, injector and column from particulate contamination from the eluent.
  • Helps to remove dissolved gases
The large surface area filters out particulate matter from the solvent that may otherwise damage expensive hardware. No tools are required for replacement.