Avantor® Hichrom, Chromatography Tubing, Precut SS
Supplier: Avantor
05-SS-10 10-SS-07 10-SS-10 10-SS-30 20-SS-10
76397-954PK 21.13 CAD
76397-954 76397-956 76397-960 76397-962 76397-964 76398-010
Avantor® Hichrom, Chromatography Tubing, Precut SS
Tubing Chromatography Tubing
316 Stainless steel, precision bore ¹/₁₆" type tubing.
  • Passivated for chemical resistance
  • Square, burr-free ends
  • Precut lengths
0.005 and 0.007" Int. Ø - suitable for minimal dead volume injector-column or column-detector connections of microbore or short columns containing 3 µm material. Sample and material must be filtered to avoid blockage.
0.010" Int. Ø - suitable for connection of analytical and semi-preparative columns containing 5 and 10 µm material. It should be used in all parts of the system where dead volume needs to be kept to minimum.
More Product Information