LiChrospher® RP-Select B, HPLC Columns
Supplier: HICHROM LIMITED
LISPRPB-5-10C5 LISPRPB-5-125A LISPRPB-5-150A LISPRPB-5-250A LISPRPB5125AF LISPRPB5250AF
76400-722EA 1503.88 CAD
76400-722 76400-724 76400-726 76400-728 76400-730 76400-732 76400-734 76400-736 76400-738 76400-740 76400-742 76400-744 76400-746 76400-748 76400-750 76400-752 76400-754 76400-756 76400-758 76400-760 76400-762 76400-764 76400-766
LiChrospher® RP-Select B, HPLC Columns
Chromatography Columns
LiChrospher® is a silica available in both endcapped and non-endcapped C8 and C18 phases.
  • Spherical porous silica
  • Base deactivated LiChrospher® RP-select B
  • High efficiency packed columns
These columns are made of stainless steel.