Avantor® Hichrom, Chromatography PTFE Tubing
Supplier: Avantor
08-PTFE-62-10M 16-PTFE-07-10M 16-PTFE-07-5M 16-PTFE-20-10M 16-PTFE-30-10M
76397-958EA 428.7 CAD
76397-958 76398-002 76398-004 76398-006 76398-008
Avantor® Hichrom, Chromatography PTFE Tubing
Tubing Chromatography Tubing
PTFE.
  • Flexible and economical
  • Excellent solvent resistance
Inert to all chemicals used in HPLC but suitable for low-pressure applications only. The most common tubing used to connect the solvent reservoir to the pump.