VeriSpec® ICP Multi-Element Internal Calibration Standard 7, RICCA Chemical Company
Supplier: Ricca Chemical
RV010858-100N RV010858-500N
RCRV010858100NEA 616.2 CAD
RCRV010858100N RCRV010858500N
VeriSpec® ICP Multi-Element Internal Calibration Standard 7, RICCA Chemical Company
Standards Cationic Standards Multi-Element Standards
Seven component standard.
10 ppm: ⁶Li, Sc, Y, In, Tb, Bi, Ge.
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility