VeriSpec® ICP Multi-Element Interference Check Solution 12, RICCA Chemical Company
Supplier: Ricca Chemical
RV010796-100N
RV010796-500N
RCRV010796100NEA
1322.46
CAD
RCRV010796100N
RCRV010796500N
VeriSpec® ICP Multi-Element Interference Check Solution 12, RICCA Chemical Company
Standards
Cationic Standards
Multi-Element Standards
12 component standard.
10000 ppm: Cl
2000 ppm: C
1000 ppm: Al, Ca, Fe, K, Mg, Na, P, S
20 ppm: Mo, Ti
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility