VeriSpec® ICP Multi-Element Instrument Calibration Standard 4, RICCA Chemical Company
Supplier: Ricca Chemical
RV010775-100N RV010775-500N
RCRV010775100NEA 597.24 CAD
RCRV010775100N RCRV010775500N
VeriSpec® ICP Multi-Element Instrument Calibration Standard 4, RICCA Chemical Company
Standards Cationic Standards Multi-Element Standards
Four component standard.
500 ppm: Ca, K, Mg, Na
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility