Deep-Freeze Color Dots for 0.2 ml PCR Tubes
Supplier: GA INTERNATIONAL INC
807656000725 807656000732 807656000749 807656000756 807656000657 807656000794 807656000664 807656000671 807656000688 807656000770 807656000701 807656000695 807656000718 807656000763 807656000787 807656024882
76274-662PK 33.56 CAD
76274-662 76274-664 76274-666 76274-668 76275-564 76275-566 76275-568 76275-570 76275-572 76275-574 76275-576 76275-578 76275-580 76275-590 76275-592 76275-594
Deep-Freeze Color Dots for 0.2 ml PCR Tubes
Labels
Color dot labels for identifying PCR tube caps, available in assorted color packs for color-coding and quick identification. These deep-freeze color dots withstand a wide temperature range, and are perfect for use throughout the lab.
  • Temperature range: −80 to +110 °C (−112 to +230 °F)
  • Dot labels designed specifically for the identification of 0.2 ml PCR tube tops and caps
  • Available in 15 colors and packs of 15 assorted colors for color-coding and easy identification
  • Labels withstand a wide temperature range; ideal for storage in laboratory refrigerators
More Product Information