Injector Support Adapter O-Ring Kit for Optima ICP-OES Instruments, PerkinElmer
Supplier: PerkinElmer
N0770438 N0770438
CA97012-754EA 100.98 CAD
CA97012-754 97012-754
Injector Support Adapter O-Ring Kit for Optima ICP-OES Instruments, PerkinElmer
Microinjectors Microinjectors, Programmable
This O-ring kit is for use with injector support adapters in Optima® 2x00/4x00/5x00/7x00 DV and 8x00 ICP-OES series instruments.
Compatible with the standard injector support adapter (97013-664) and the injector support adapter for 1.2 mm I.D. entire length injectors (101688-042).
Delivery information: Includes O-ring 8.74 mm I.D (2), O-ring 6.07 mm I.D (2), washer (2).