HOBO® Water Level Data Logger, Bluetooth-Enabled
Supplier: ONSET COMPUTER CORPORATION SE
MX2001-01-SS MX2001-01-TI MX2001-02-SS MX2001-03-SS MX2001-04-SS MX2001-04-TI CABLE-RWL-010 CABLE-RWL-015 CABLE-RWL-030 CABLE-RWL-5.0
470323-356EA 1743.9 CAD
470323-356 470323-360 470323-364 470323-368 470323-372 470323-376 470324-406 470324-408 470324-410 470324-416
HOBO® Water Level Data Logger, Bluetooth-Enabled
Data Loggers
Wireless setup and download via bluetooth technology. Lowers the cost of field data collection by providing wireless access to high-accuracy water level and temperature data.
  • Wireless data offload to mobile devices or windows computers via bluetooth
  • Integrated barometric pressure sensor enables direct water level readout
  • Available with stainless steel or titanium sensor ends
  • 3-point NIST-traceable calibration certificate included for the water pressure sensor
Consists of a top-end unit and a water level sensor, which are sold as a set, plus a direct-More Product Information