WHEATON® Polypropylene Screw Caps, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
W240848 W240849 W240851
10796-632CS 551.51 CAD
10796-632 10796-634 10796-638
WHEATON® Polypropylene Screw Caps, DWK Life Sciences
Closures Bottle Closures
These caps for use with premium quality WHEATON® glass bottles or vials.
  • Polyvinyl liners can be used with mild acids, alkalis, solvents, and alcohols
  • Foamed polyethylene (PE) liner provides chemical resistance for acids, alkalis, solvents, and alcohols
  • PTFE faced foamed PE liner ideal for use with high purity chemicals,strong acids, and solvents
  • Open top caps provide access with a syringe
  • Open top caps with pre-slit liner are ideal for use in automated liquidhandling equipment
WHEATON® offers a wide variety of caps and closures to meet every unique need.