WHEATON® Rubber Stoppers, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
W224100-282 224100-174 W224100-342
89427-454CS 1586.03 CAD
89427-454 10028-042 10796-550
WHEATON® Rubber Stoppers, DWK Life Sciences
Stoppers Rubber Stoppers
Clean and precise pharmaceutical grade rubber stoppers for research/parenteral drug applications.
  • Variety of styles, sizes and rubber formulations
  • Components manufactured to yield exceptional dimensional stability
  • Straight plug stoppers provide maximum sealing for vials with straight-wall glass finish
  • Snap-on style provides maximum sealing for vials with a blow-back glass finish
  • Sleeve style stoppers feature a fold-over skirt for sealing in serum finish vials without a crimp closure
  • Autoclavable
WHEATON® straight plug stoppers are manufactured from the highest quality raw materials.