WHEATON® Erlenmeyer Shake Flasks with DuoCap® Closures, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
WPFBC0500S WPFBC1000S WPFBC2000S WPFBC3000S
10017-888CS 342.58 CAD
10017-888 10017-890 10017-892 10017-894
WHEATON® Erlenmeyer Shake Flasks with DuoCap® Closures, DWK Life Sciences
Flasks Erlenmeyer Flasks
WHEATON® offers both flat bottom and baffled Erlenmeyer flasks.
  • Ideal replacement for glass and other Erlenmeyer/shaker flasks
  • Patented DuoCap® allows sterile air exchange or a leak-resistant seal
  • Minimize oxidation or pH shifts
  • Perform aerobic/anaerobic culture
  • Available with septum cap for pathogen containment
  • Re-autoclavable, reusable
  • Individually packed in easy tear bags
These Erlenmeyer flasks are made of Lexan polycarbonate resin, which offers outstanding mechanical, optical, electrical, and thermal properties. This resin conforms to flameMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable