Toluene ACS

Supplier: Ricca Chemical
Danger

Synonyms: Methylbenzene, Phenylmethane, FM approval

RSOT0200-4C RSOT0200-20E4
RCRSOT0200-4CEA 353.92 CAD
RCRSOT0200-4C RCRSOT020020E4 RCRSOT02004X4C
Toluene ACS
Toluene
Formula: C₆H₅CH₃
MW: 92.14 g/mol
Boiling Pt: 110.6
Melting Pt: -95
Density: 0.8660
Flash Pt: 4°C
MDL Number: MFCD00008512
CAS Number: 108-88-3
UN: 1294
ADR: 3 II
Merck Index: 13,09607
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=44efa31292a7dce6d9e60bcbaec208eb42d34e32&vid=73ada9ad1e894808b6c193cabc4babcfaf320a25&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay min 99.5 %
Color (APHA) max 10
Sulfur compounds (as S) max 0.003 %
Water (H₂O) max 0.03 %
Residue after evaporation (Non-Volatile Matter) max 0.001 %
Substances Darkened by Sulfuric Acid To Pass Test

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR