Chloroform ACS

Supplier: Ricca Chemical
Danger

RSOC0020-1C RSOC0020-4C RSOC0020-500C
RCRSOC0020-1CEA 151.68 CAD
RCRSOC0020-1C RCRSOC0020-4C RCRSOC0020500C
Chloroform ACS
Chloroform
Formula: CHCl₃
Boiling Pt: 60.5…61.2°C
Melting Pt: –63.5°C
Density: 1.4840
MDL Number: MFCD00000826
CAS Number: 67-66-3
UN: 1888
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9cc00da8cefd4e04c0ea1d7c5801308cd73eeef9&vid=c7d46a2ee73651eae0f3654c272e820106b2f04f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay min 99.8 %
Color (APHA) max 10
Acetone and Aldehyde To Pass Test
Free Chlorine (Cl) To Pass Test
Lead (Pb) max 0.05 ppm
Acid and Chloride To Pass Test
Residue after evaporation (Non-Volatile Matter) max 0.001 %
Substances Darkened by Sulfuric Acid To Pass Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR