Acetone ACS

Supplier: Ricca Chemical
Danger

Synonyms: 2-Propanone, DMK, Propanone, Dimethyl ketone

RSOA0010-1C RSOA0010-20F RSOA0010-4A RSOA0010-4C RSOA0010-20E4 RSOA0010-500C
RCRSOA0010-1CEA 121.66 CAD
RCRSOA0010-1C RCRSOA0010-20F RCRSOA0010-4A RCRSOA0010-4C RCRSOA0010-55E RCRSOA001020E4 RCRSOA00104X4A RCRSOA00104X4C RCRSOA0010500C
Acetone ACS
Acetone
Formula: CH₃COCH₃
MW: 58.08 g/mol
Boiling Pt: 56.05°C (1013.25 hPa)
Melting Pt: –94.7°C
Density: 0.7880
Flash Pt: –20°C
MDL Number: MFCD00008765
CAS Number: 67-64-1
UN: 1090
ADR: 3 II
Merck Index: 13,00067
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3de5d8e3bf7a94692d684c8f0ccdfb8500fde76d&vid=24fb2e0ec86974b1803780c7d5b23804a085df64&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay min 99.5 %
Color (APHA) max 10
Solubility in Water To Pass Test
Isopropyl Alcohol max 0.05 %
Methanol (Methyl Alcohol) max 0.05 %
Water (H₂O) max 0.5 %
Aldehyde (as HCHO) max 0.002 %
Residue after evaporation (Non-Volatile Matter) max 0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR