Acetic acid glacial ACS
Supplier: Ricca Chemical
|
Danger
|
RABA0010-1C
RABA0010-4C
RABA0010-20E1
RABA0010-250A
RABA0010-2.5D1
RABA0010-500D1
RCRABA0010-1CEA
282.82
CAD
RCRABA0010-1C
RCRABA0010-4C
RCRABA001020E1
RCRABA0010250A
RCRABA001025D1
RCRABA00104X25
RCRABA010500D1
Acetic acid glacial ACS
Acetic acid
Formula:
H₃CCOOH Boiling Pt: ∼ 118°C Melting Pt: 16.64°C Density: 1.0496 Flash Pt: 39°C |
MDL Number:
MFCD00036152 CAS Number: 64-19-7 UN: 2789 ADR: 8 (3),II |
|
Specification Test Results
Assay | min 99.7 % |
Color (APHA) | max 10 |
Dilution test | To Pass Test |
Acetic Anhydride [(CH₃CO)₂O] | max 0.01 % |
Heavy Metals (as Pb) | max 0.5 ppm |
Sulfate (SO₄) | max 1 ppm |
Chloride (Cl) | max 1 ppm |
Iron (Fe) | max 0.2 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...