Tetrabutylammonium hydroxide 0.100 Normal in methanol
Supplier: Ricca Chemical
|
Danger
|
R8384000-1C
R8384000-4C
RCR8384000-1CEA
1565.78
CAD
RCR8384000-1C
RCR8384000-4C
Tetrabutylammonium hydroxide 0.100 Normal in methanol
Tetrabutylammonium hydroxide
Formula:
C₁₆H₃₇NO Boiling Pt: ∼ 64.5°C Density: 0.8000 Flash Pt: ∼ 11°C |
MDL Number:
MFCD00009425 CAS Number: 2052-49-5 UN: 1230 ADR: 3,II |
Specification Test Results
Appearance | Colorless to slightly yellow liquid |
Assay (vs. Sulfuric Acid/Phenolphthalein) | 0.0995-0.1005 N at 20°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...