2-Propanol 70% (v/v)
Supplier: Ricca Chemical
|
Danger
|
R4210000-55E
R4210000-20E1
R4210000-20F
R4210000-4A
R4210000-4C1
RC4210-55EA
4084.3
CAD
RC4210-55
RC4210-5HP
RC42105
RC42101
RCR4210000-4C1
2-Propanol 70% (v/v)
2-Propanol
Formula:
(CH₃)₂CHOH Boiling Pt: ∼ 82°C Melting Pt: ∼ 0°C Density: 0.8770 Flash Pt: ∼ 13°C (55°F) (100%); 18°C (64°F) (70%) |
MDL Number:
MFCD00011674 CAS Number: 67-63-0 UN: 1219 ADR: 3,II |
|
Specification Test Results
Appearance | Colorless liquid |
Assay (by Specific Gravity) | 68-72 % (v/v) |
Specific Gravity at 20°C | 0.874-0.882 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...