Sodium hydrogen carbonate saturated
Supplier: Ricca Chemical
7148-1
7148-16
7148-32
7148-5
RC7148-1EA
135.88
CAD
RC7148-1
RC7148-16
RC7148-32
RC7148-5
Sodium hydrogen carbonate saturated
Sodium hydrogen carbonate
Formula:
NaHCO₃ Boiling Pt: ∼ 100°C Melting Pt: ∼ 0°C Density: 1.0700 |
MDL Number:
MFCD00003528 CAS Number: 144-55-8 |
|
Specification Test Results
Appearance | Colorless liquid |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...