Copper (II) ethylenediamine complex 1 Molar

Supplier: Ricca Chemical
Danger

Synonyms: Bis(ethylenediamine)copper dihydroxide, Cupriethylendiamine, Copper(II)-ethylenediame complex

2499-1 2499-32
RC2499-1EA 788.42 CAD
RC2499-1 RC2499-32
Copper (II) ethylenediamine complex 1 Molar
Copper (II) ethylenediamine
Formula: Cu(H₂NCH₂CH₂NH₂)₂(OH)₂
MW: 217.76 g/mol
Density: 1.1000
MDL Number: MFCD00274628
CAS Number: 14552-35-3
UN: 1761
ADR: 8 (6.1) III

Order Now

Specification Test Results

Appearance blue-purple liquid
Assay (vs. Sodium Thiosulfate/Starch) 0.98-1.02 Molar
Ethylenediamine / Cupric Ratio 1.96-2.04


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR