Labels 'Hazardous Waste', Labelmaster
Supplier: Label Master
HWA HWAV HWAV500
101376-284EA 88.05 CAD
101376-284 101377-376 101377-374
Labels 'Hazardous Waste', Labelmaster
Labels
Avoid potential dangers and stay in regulatory compliance with Hazardous Waste labels.
  • Constructed of paper
  • Economical alternative to vinyl
These labels provide a convenient way for a generator to mark accumulation containers in accordance with EPA regulations in 40 CFR part 262.34.