VWR® P-Series Balances with Calibration Certificate
Supplier: VWR International
VWR-123P/C VWR-500P/C VWR-1500P/C VWR-3000P/C VWR-6000P/C
76378-352EA 1383.52 CAD
76378-352 76378-354 76378-356 76378-358 76378-360
VWR® P-Series Balances with Calibration Certificate
Balances and Scales
The VWR® P-Series/C balances includes a certificate of calibration showing the full testing data from an accredited Calibration and Testing Laboratory utilizing NIST-traceable weights are the answer for an organizations regulatory and compliance needs.
  • Complete calibration data reported
  • Portable design
  • Stainless steel weighing pan
  • Rugged construction
  • Five years warranty
These economical, portable balances are ideal for educational, light industrial, and general laboratory applications. These portable balances offer simple operation and multiple weighing modes.
The model with 0.001 g readability features a removable draft shield. The models with 0.01 g readability and below include a plastic weighing bowl.
Certifications: ISO/IEC 17025:2017 accredited Calibration and Testing Laboratory utilizing NIST-traceable weights, CE marked.
Delivery information: All units are supplied with power adapter, weighing pan, and user manual. Batteries are not included.