KIMBLE® MICROFLEX® Kjeldal-Shaped Flasks, Threaded, DWK Life Sciences
Supplier: DWK Life Sciences
KIMBLE®
747550-1020 747550-2520
KT747550-1020EA 66.58 CAD
KT747550-1020 KT747550-2520
KIMBLE® MICROFLEX® Kjeldal-Shaped Flasks, Threaded, DWK Life Sciences
Flasks Kjeldahl Flasks
This flask has a conventional round bottom shape with a tapered upper portion for easy recovery of reaction products.
  • Accessory for Microscale MICROFLEX® Standard Taper 14/10 threaded kits
Manufactured from 33 expansion, low extractable borosilicate glass.
Certifications: Glass conforms to USP Type I and ASTM E438, Type I, Class A requirements.