Ward's® Mini LED White-Light Box
Supplier: Ward's
Ward's®
120661W
470311-200CS 5533.6 CAD
470311-200
Ward's® Mini LED White-Light Box
Lights Light Boxes
This mini light box saves bench space and can be easily stored in a drawer.
  • LED light source stays cool and has a longer life (10000 hours)
  • Battery-operated
  • Great for use with the Wards® mini magnifier (available separately)
The viewing area on this light box is is perfect for quick screening of 96-well plates, petri dishes, electrophoresis gels, slides, and transparencies.
Delivery information: Includes six AAA batteries.